missing translation for 'onlineSavingsMsg'
Learn More

3-(Trifluoromethyl)benzoic acid, 98+%

Product Code. 11452748
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11452748 10 g 10g
11462748 50 g 50g
11329549 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11452748 Supplier Thermo Scientific Alfa Aesar Supplier No. A16737.09
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 454-92-2 | C8H5F3O2 | 190.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 454-92-2
Molecular Formula C8H5F3O2
Molecular Weight (g/mol) 190.12
MDL Number MFCD00002519
InChI Key FQXQBFUUVCDIRK-UHFFFAOYSA-N
Synonym 3-trifluoromethyl benzoic acid, m-trifluoromethylbenzoic acid, alpha,alpha,alpha-trifluoro-m-toluic acid, 3-carboxybenzotrifluoride, 3-trifluoromethyl-benzoic acid, benzoic acid, 3-trifluoromethyl, m-trifluoromethyl benzoic acid, 3-trifluoromethyl benzoicacid, m-carboxybenzotrifluoride, a,a,a-trifluoro-m-toluic acid
PubChem CID 9963
ChEBI CHEBI:60695
IUPAC Name 3-(trifluoromethyl)benzoic acid
SMILES OC(=O)C1=CC=CC(=C1)C(F)(F)F

Specifications

Melting Point 103°C to 107°C
Boiling Point 237°C to 240°C
Quantity 10 g
Beilstein 2049239
Formula Weight 190.12
Percent Purity ≥98%
Chemical Name or Material 3-(Trifluoromethyl)benzoic acid
compliance-icons
Product Identifier
  • 3-(Trifluoromethyl)benzoic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: alpha.,.alpha.,.alpha.-Trifluoro-m-toluic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.