missing translation for 'onlineSavingsMsg'
Learn More

3-(Trifluoromethyl)benzoic acid, 99%, Thermo Scientific™

Product Code. 10427200 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10427200 10 g Glass bottle 10g
10592201 50 g Glass bottle 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10427200 Supplier Thermo Scientific Acros Supplier No. 182540100

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 454-92-2
Molecular Formula C8H5F3O2
Molecular Weight (g/mol) 190.12
MDL Number MFCD00002519
InChI Key FQXQBFUUVCDIRK-UHFFFAOYSA-N
Synonym 3-trifluoromethyl benzoic acid, m-trifluoromethylbenzoic acid, alpha,alpha,alpha-trifluoro-m-toluic acid, 3-carboxybenzotrifluoride, 3-trifluoromethyl-benzoic acid, benzoic acid, 3-trifluoromethyl, m-trifluoromethyl benzoic acid, 3-trifluoromethyl benzoicacid, m-carboxybenzotrifluoride, a,a,a-trifluoro-m-toluic acid
PubChem CID 9963
ChEBI CHEBI:60695
IUPAC Name 3-(trifluoromethyl)benzoic acid
SMILES OC(=O)C1=CC=CC(=C1)C(F)(F)F

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 104°C to 106°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula CF3C6H4CO2H
Quantity 10 g
Beilstein 09, 478
Formula Weight 190.12
Percent Purity ≥98.5%
Physical Form Powder
Chemical Name or Material 3-(Trifluoromethyl)benzoic acid, 99%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.