missing translation for 'onlineSavingsMsg'
Learn More

3-(Trifluoromethyl)benzoyl chloride, 97%

Product Code. 11451828
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11451828 5 g 5g
11461828 25 g 25g
11471828 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11451828 Supplier Thermo Scientific Alfa Aesar Supplier No. A14493.06
 Restricted Item
Estimated Shipment: 10-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 2251-65-2 | C8H4ClF3O | 208.564 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2251-65-2
Molecular Formula C8H4ClF3O
Molecular Weight (g/mol) 208.564
MDL Number MFCD00000680
InChI Key RUJYJCANMOTJMO-UHFFFAOYSA-N
Synonym 3-trifluoromethyl benzoyl chloride, benzoyl chloride, 3-trifluoromethyl, m-trifluoromethyl benzoyl chloride, m-trifluoromethylbenzoyl chloride, alpha,alpha,alpha-trifluoro-m-toluoyl chloride, 3-trifluoromethyl benzoylchloride, 3-trifluoromethyl-benzoyl chloride, alpha,alpha,alpha-trifluoro-m-tolyl chloride, pubchem5468
PubChem CID 75257
IUPAC Name 3-(trifluoromethyl)benzoyl chloride
SMILES C1=CC(=CC(=C1)C(F)(F)F)C(=O)Cl

Specifications

Density 1.408
Boiling Point 189°C to 191°C
Flash Point >110°C (230°F)
Odor Pungent
Refractive Index 1.477
Quantity 5 g
UN Number UN3265
Beilstein 391266
Sensitivity Moisture sensitive
Formula Weight 208.57
Percent Purity 97%
Chemical Name or Material 3-(Trifluoromethyl)benzoyl chloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.