missing translation for 'onlineSavingsMsg'
Learn More

Deoxy Modafinil, TRC

Product Code. 30359905
Change view
Click to view available options
Quantity:
50 mg
250 mg
1 g
Unit Size:
1g
250mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30359905 250 mg 250mg
30351789 50 mg 50mg
30363278 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30359905 Supplier Toronto Research Chemicals Supplier No. TRCD242700250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 68524-30-1
Molecular Formula C15 H15 N O S
Molecular Weight (g/mol) 257.35
Synonym 2-[(Diphenylmethyl)sulfenyl]acetamide
IUPAC Name 2-benzhydrylsulfanylacetamide
SMILES NC(=O)CSC(c1ccccc1)c2ccccc2

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C15H15NOS/c16-14(17)11-18-15(12-7-3-1-4-8-12)13-9-5-2-6-10-13/h1-10,15H,11H2,(H2,16,17)
Formula Weight 257.0874
Chemical Name or Material 2-[(Diphenylmethyl)sulfenyl]acetamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.