missing translation for 'onlineSavingsMsg'
Learn More

(2,3-Dimethylphenyl)[1-(trityl)-1H-imidazol-4-yl]methanone, TRC

Product Code. 30374134
Change view
Click to view available options
Quantity:
25 mg
100 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30374134 250 mg
30385242 100 mg
30382903 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30374134 Supplier Toronto Research Chemicals Supplier No. TRCD477350250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 176721-02-1
Molecular Formula C31H26N2O
Molecular Weight (g/mol) 442.55
Synonym (2,3-Dimethylphenyl)[1-(triphenylmethyl)-1H-imidazol-4-yl]methanone
SMILES Cc1cccc(C(=O)c2cn(cn2)C(c3ccccc3)(c4ccccc4)c5ccccc5)c1C

Specifications

Quantity 250 mg
Formula Weight 442.2045
Chemical Name or Material (2,3-Dimethylphenyl)[1-(trityl)-1H-imidazol-4-yl]methanone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.