missing translation for 'onlineSavingsMsg'
Learn More

5-Phenyl-3-isoxazolyl Sulfate Potassium, TRC

Product Code. 30413880
Change view
Click to view available options
Quantity:
25 mg
250 mg
Unit Size:
250mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30413880 25 mg 25mg
30414854 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30413880 Supplier Toronto Research Chemicals Supplier No. TRCP33547025MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 5-Phenyl-3-isoxazolyl Sulfate Potassium
Analyte or Component Names 5-Phenyl-3-isoxazolyl Sulfate Potassium
Shipping Condition Dry ice
Molecular Formula C9 H6 K N O5 S
Synonym 5-Phenyl-3-isoxazolol 3-(Hydrogen Sulfate) Potassium, 5-Phenyl-3-isoxazolol Hydrogen Sulfate (Ester) Potassium
InChI Formula InChI=1S/C9H7NO5S.K/c11-16(12,13)15-9-6-8(14-10-9)7-4-2-1-3-5-7;/h1-6H,(H,11,12,13);/q;+1/p-1
Solution Type Neat
SMILES [K]OS(=O)(=O)Oc1cc(on1)c2ccccc2
IUPAC Name (5-phenyl-1,2-oxazol-3-yl)oxysulfonyloxypotassium
Industry Type Pharmaceutical
Molecular Weight (g/mol) 279.311
Quantity 25 mg
Formula Weight 278.96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.