missing translation for 'onlineSavingsMsg'
Learn More

N-Nitroso Desmethyl Sumatriptan, TRC

Product Code. 30414092
Change view
Click to view available options
Quantity:
2.5 mg
10 mg
25 mg
Unit Size:
10mg
2.5mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30414092 10 mg 10mg
30426318 2.5 mg 2.5mg
30409486 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30414092 Supplier Toronto Research Chemicals Supplier No. TRCN79182010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-Nitroso Desmethyl Sumatriptan
Analyte or Component Names N-Nitroso Desmethyl Sumatriptan
Purity Grade Notes HPLC
Shipping Condition Room Temperature
Molecular Formula C13 H18 N4 O3 S
InChI Formula InChI=1S/C13H18N4O3S/c1-14-21(19,20)9-10-3-4-13-12(7-10)11(8-15-13)5-6-17(2)16-18/h3-4,7-8,14-15H,5-6,9H2,1-2H3
Solution Type Neat
SMILES CNS(=O)(=O)Cc1ccc2[nH]cc(CCN(C)N=O)c2c1
IUPAC Name N-methyl-1-[3-[2-[methyl(nitroso)amino]ethyl]-1H-indol-5-yl]methanesulfonamide
Industry Type Pharmaceutical
Molecular Weight (g/mol) 310.372
Quantity 10 mg
Formula Weight 310.11
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.