missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Suberoylanilide Hydroxamic Acid Beta-D-Glucuronide Sodium Salt, TRC
Specifications
Specifications
| Chemical Name or Material | Suberoylanilide Hydroxamic Acid beta-D-Glucuronide Sodium Salt |
| Analyte or Component Names | Suberoylanilide Hydroxamic Acid beta-D-Glucuronide Sodium Salt |
| Shipping Condition | Room Temperature |
| Molecular Formula | C20H27N2NaO9 |
| Synonym | 1-O-[[1,8-Dioxo-8-(phenylamino)octyl]amino]-β-D-glucopyranuronic Acid Sodium Salt, Vorinostat glucuronide Sodium Salt |
| InChI Formula | InChI=1S/C4H6N4O/c5-8-3-1-4(9)7-2-6-3/h1-2H,5H2,(H2,6,7,8,9) |
| Solution Type | Neat |
| SMILES | NNc1cc(O)ncn1 |
| Industry Type | Pharmaceutical |
| Molecular Weight (g/mol) | 462.43 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?