missing translation for 'onlineSavingsMsg'
Learn More

SEPHS1 Antibody [DyLight 755], Novus Biologicals Biologicals™

Product Code. 30487225 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30487225 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30487225 Supplier Novus Biologicals Supplier No. NBP335339IR

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

SEPHS1 Polyclonal antibody specifically detects SEPHS1 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot,Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen SEPHS1
Applications ELISA, Western Blot, Immunocytochemistry/Immunofluorescence
Classification Polyclonal
Conjugate DyLight 755
Formulation 50mM Sodium Borate
Gene Alias EC 2.7.9.3, MGC4980, SELD, Selenium donor protein 1, Selenophosphate synthase 1, selenophosphate synthetase 1, selenophosphate synthetase 1 +E9a, selenophosphate synthetase 1 delta E2, selenophosphate synthetase 1 delta E8, selenophosphate synthetase 1 major, SPS1selenophosphate synthetase 1 +E9, water dikinase 1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-392 of human SEPHS1 (NP_036379.2).,, Sequence:, MSTRESFNPESYELDKSFRLTRFTELKGTGCKVPQDVLQKLLESLQENHFQEDEQFLGAVMPRLGIGMDTCVIPLRHGGLSLVQTTDYIYPIVDDPYMMGRIACANVLSDLYAMGVTECDNMLMLLGVSNKMTDRERDKVMPLIIQGFKDAAEEAGTSVTGGQTVLNPWIVLGGVATTVCQPNEFIMPDNAVPGDVLVLTKPLGTQVAVAVHQWLDIPEKWNKIKLVVTQEDVELAYQEAMMNMARLNRTAAGLMHTFNAHAATDITGFGILGHAQNLAKQQRNEVSFVIHNLPVLAKMAAVSKACGNMFGLMHGTCPETSGGLLICLPREQAARFCAEIKSPKYGEGHQAWIIGIVEKGNRTARIIDKPRIIEVAPQVATQNVNPTPGATS
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 22929
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.