missing translation for 'onlineSavingsMsg'
Learn More

ULK2 Antibody [CoraFluorÖ 1], Novus Biologicals Biologicals™

Product Code. 30491355 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30491355 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30491355 Supplier Novus Biologicals Supplier No. NBP327885CL1

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

ULK2 Polyclonal antibody specifically detects ULK2 in Human samples. It is validated for ELISA,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen ULK2
Applications ELISA, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence
Classification Polyclonal
Conjugate CoraFluor 1
Formulation PBS
Gene Alias ATG1B, EC 2.7.11, EC 2.7.11.1, KIAA0623serine/threonine-protein kinase ULK2, unc-51 (C. elegans)-like kinase 2, Unc51.2, Unc-51-like kinase 2, unc-51-like kinase 2 (C. elegans)
Host Species Rabbit
Immunogen ULK2 antibody was raised against a 17 amino acid peptide near the center of human ULK2 . The immunogen is located within amino acids 480 - 530 of ULK2.
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 9706
Target Species Human
Content And Storage Store at 4°C in the dark. Do not freeze.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.