missing translation for 'onlineSavingsMsg'
Learn More

RASSF1 Antibody [mFluor Violet 450 SE], Novus Biologicals Biologicals™

Product Code. 30493743 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30493743 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30493743 Supplier Novus Biologicals Supplier No. NBP335204MFV450

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

RASSF1 Polyclonal antibody specifically detects RASSF1 in Human samples. It is validated for ELISA,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen RASSF1
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate mFluor Violet 450 SE
Formulation 50mM Sodium Borate
Gene Alias 123F2, NORE2A, pancreas-specific ras association domain family 1 protein, Ras association (RalGDS/AF-6) domain family member 1, ras association domain-containing protein 1, RASSF1A, RDA32cardiac-specific ras association domain family 1 protein, REH3P21, tumor suppressor protein RDA32, WUGSC:H_LUCA12.5
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 101-340 of human RASSF1 (NP_009113.3).,, Sequence:, CCGPRDLGWEPAVERDTNVDEPVEWETPDLSQAEIEQKIKEYNAQINSNLFMSLNKDGSYTGFIKVQLKLVRPVSVPSSKKPPSLQDARRGPGRGTSVRRRTSFYLPKDAVKHLHVLSRTRAREVIEALLRKFLVVDDPRKFALFERAERHGQVYLRKLLDDEQPLRLRLLAGPSDKALSFVLKENDSGEVNWDAFSMPELHNFLRILQREEEEHLRQILQKYSYCRQKIQEALHACPLG
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Breast Cancer, Cell Cycle and Replication, Mitotic Regulators
Primary or Secondary Primary
Gene ID (Entrez) 11186
Target Species Human
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.