missing translation for 'onlineSavingsMsg'
Learn More

TUG Antibody (4A11A6G11) [Alexa Fluor« 532], Novus Biologicals Biologicals™

Product Code. 30501701 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30501701 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30501701 Supplier Novus Biologicals Supplier No. NBP327915AF532

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

TUG Monoclonal antibody specifically detects TUG in Mouse samples. It is validated for ELISA,Western Blot,Serum
TRUSTED_SUSTAINABILITY

Specifications

Antigen TUG
Applications ELISA, Western Blot
Classification Monoclonal
Clone 4A11A6G11
Conjugate Alexa Fluor 532
Formulation 50mM Sodium Borate
Gene Alias Alveolar soft part sarcoma chromosomal region candidate gene 1 protein, alveolar soft part sarcoma chromosome region, candidate 1, Alveolar soft part sarcoma locus, ASPLUBX domain-containing protein 9, ASPSrenal cell carcinoma gene on chromosome 17, RCC17renal cell carcinoma, papillary, 17, Renal papillary cell carcinoma protein 17, tether containing UBX domain for GLUT4, TUG, UBX domain protein 9, UBXD9ASPCR1, UBXN9FLJ45380
Host Species Mouse
Immunogen Truncated recombinant TUG expressed in E. Coli strain BL21 (DE3).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cell Biology, Endocrinology, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 79058
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.