missing translation for 'onlineSavingsMsg'
Learn More

COG2 Antibody [DyLight 488], Novus Biologicals Biologicals™

Product Code. 30507112 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30507112 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30507112 Supplier Novus Biologicals Supplier No. NBP338235G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

COG2 Polyclonal antibody specifically detects COG2 in Human,Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen COG2
Applications ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 488
Formulation 50mM Sodium Borate
Gene Alias component of oligomeric golgi complex 2COG complex subunit 2, conserved oligomeric Golgi complex protein 2, conserved oligomeric Golgi complex subunit 2, LDLCbrefeldin A-sensitive, peripheral Golgi protein, low density lipoprotein receptor defect C complementing, Low density lipoprotein receptor defect C-complementing protein
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-280 of human COG2 (NP_031383.1).,, Sequence:, MEKSRMNLPKGPDTLCFDKDEFMKEDFDVDHFVSDCRKRVQLEELRDDLELYYKLLKTAMVELINKDYADFVNLSTNLVGMDKALNQLSVPLGQLREEVLSLRSSVSEGIRAVDERMSKQEDIRKKKMCVLRLIQVIRSVEKIEKILNSQSSKETSALEASSPLLTGQILERIATEFNQLQFHAVQSKGMPLLDKVRPRIAGITAMLQQSLEGLLLEGLQTSDVDIIRHCLRTYATIDKTRDAEALVGQVLVKPYIDEVIIEQFVESHPNGLQVMYNKLL
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Golgi Apparatus Markers
Primary or Secondary Primary
Gene ID (Entrez) 22796
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.