missing translation for 'onlineSavingsMsg'
Learn More

AP180 Antibody [mFluor Violet 450 SE], Novus Biologicals Biologicals™

Product Code. 30509759 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30509759 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30509759 Supplier Novus Biologicals Supplier No. NBP338325MFV450

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

AP180 Polyclonal antibody specifically detects AP180 in Mouse,Rat samples. It is validated for ELISA,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen AP180
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate mFluor Violet 450 SE
Formulation 50mM Sodium Borate
Gene Alias 91 kDa synaptosomal-associated protein, AP180, CALM, clathrin coat assembly protein AP180, DKFZp781O0519, KIAA0656synaptosomal-associated protein, 91 kDa (mouse) homolog, Phosphoprotein F1-20, synaptosomal-associated protein, 91kDa homolog (mouse)
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 321-600 of human AP180 (NP_001229723.1).,, Sequence:, AVPVSTSKPSSDLLDLQPDFSSGGAAAAAAPAPPPPAGGATAWGGFGGSFMAPSPSPVTPAQNNLLQPNFEAAFGTTPSTSSSSSFDPSVFDGLGDLLMPTMAPAGQPAPVSMVPPSPAMAASKALGSDLDSSLASLVGNLGISGTTTKKGDLQWNAGEKKLTGGANWQPKVAPATWSAGVPPSAPLQGAVPPTSSVPPVAGAPSVGQPGAGFGMPPAGTGMPMMPQQPVMFAQPMMRPPFGAAAVPGTQLSPSPTPASQSPKKPPAKDPLADLNIKDFL
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9892
Target Species Mouse, Rat
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.