missing translation for 'onlineSavingsMsg'
Learn More

FANCL Antibody [DyLight 650], Novus Biologicals Biologicals™

Product Code. 30510107 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30510107 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30510107 Supplier Novus Biologicals Supplier No. NBP338338C

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

FANCL Polyclonal antibody specifically detects FANCL in Mouse samples. It is validated for ELISA,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen FANCL
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate DyLight 650
Formulation 50mM Sodium Borate
Gene Alias E3 ubiquitin-protein ligase FANCL, EC 6.3.2.-, FAAP43PHF9FLJ10335, Fanconi anemia group L protein, Fanconi anemia, complementation group L, Fanconi anemia-associated polypeptide of 43 kDa, PHD finger protein 9, Pog
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-255 of human FANCL (NP_060532.2).,, Sequence:, MAVTEASLLRQCPLLLPQNRSKTVYEGFISAQGRDFHLRIVLPEDLQLKNARLLCSWQLRTILSGYHRIVQQRMQHSPDLMSFMMELKMLLEVALKNRQELYALPPPPQFYSSLIEEIGTLGWDKLVYADTCFSTIKLKAEDASGREHLITLKLKAKYPAESPDYFVDFPVPFCASWTPQSSLISIYSQFLAAIESLKAFWDVMDEIDEKTWVLEPEKPPRSATARRIALGNNVSINIEVDPRHPTMLPECFFLG
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, DNA Repair, Genes Sensitive to DNA Damaging Agents
Primary or Secondary Primary
Gene ID (Entrez) 55120
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.