missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
APIP Polyclonal antibody specifically detects APIP in Human,Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
Specifications
Specifications
| Antigen | APIP |
| Applications | ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Alexa Fluor 488 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | APAF1 interacting protein, APAF1-interacting protein, APIP2, CGI29, CGI-29, dJ179L10.2, EC 4.2.1.109, MMRP19, MTRu-1-P dehydratase, probable methylthioribulose-1-phosphate dehydratase |
| Host Species | Rabbit |
| Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1-242 of human APIP (NP_057041.2).,, Sequence:, MSGCDAREGDCCSRRCGAQDKEHPRYLIPELCKQFYHLGWVTGTGGGISLKHGDEIYIAPSGVQKERIQPEDMFVCDINEKDISGPSPSKKLKKSQCTPLFMNAYTMRGAGAVIHTHSKAAVMATLLFPGREFKITHQEMIKGIKKCTSGGYYRYDDMLVVPIIENTPEEKDLKDRMAHAMNEYPDSCAVLVRRHGVYVWGETWEKAKTMCECYDYLFDIAVSMKKVGLDPSQLPVGENGIV |
| Purification Method | Affinity purified |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?