missing translation for 'onlineSavingsMsg'
Learn More

SMCHD1 Antibody [Alexa Fluor« 647], Novus Biologicals Biologicals™

Product Code. 30511842 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30511842 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30511842 Supplier Novus Biologicals Supplier No. NBP338399AF647

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

SMCHD1 Polyclonal antibody specifically detects SMCHD1 in Human,Rat samples. It is validated for ELISA,Western Blot,Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen SMCHD1
Applications ELISA, Western Blot, Immunocytochemistry/Immunofluorescence
Classification Polyclonal
Conjugate Alexa Fluor 647
Formulation 50mM Sodium Borate
Gene Alias DKFZp686O0631, KIAA0650, structural maintenance of chromosomes flexible hinge domain containing 1, structural maintenance of chromosomes flexible hinge domain-containing protein 1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1756-2005 of human SMCHD1 (NP_056110.2).,, Sequence:, TTDAARRIYDETQGRQQVLPLDSIYKKTLPDWKRSLPHFRNGKLYFKPIGDPVFARDLLTFPDNVEHCETVFGMLLGDTIILDNLDAANHYRKEVVKITHCPTLLTRDGDRIRSNGKFGGLQNKAPPMDKLRGMVFGAPVPKQCLILGEQIDLLQQYRSAVCKLDSVNKDLNSQLEYLRTPDMRKKKQELDEHEKNLKLIEEKLGMTPIRKCNDSLRHSPKVETTDCPVPPKRMRREATRQNRIITKTDV
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 23347
Target Species Human, Rat
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.