missing translation for 'onlineSavingsMsg'
Learn More

PDCD2L Antibody [DyLight 755], Novus Biologicals Biologicals™

Product Code. 30512574 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30512574 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30512574 Supplier Novus Biologicals Supplier No. NBP338423IR

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

PDCD2L Polyclonal antibody specifically detects PDCD2L in Human,Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen PDCD2L
Applications ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 755
Formulation 50mM Sodium Borate
Gene Alias MGC13096, programmed cell death 2-like, programmed cell death protein 2-like
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-358 of human PDCD2L (NP_115722.1).,, Sequence:, MAAVLKPVLLGLRDAPVHGSPTGPGAWTASKLGGIPDALPTVAAPRPVCQRCGQPLALVVQVYCPLEGSPFHRLLHVFACACPGCSTGGARSWKVFRSQCLQVPEREAQDAQKQGNSLAAEDWCEGADDWGSDTEEGPSPQFTLDFGNDASSAKDVDWTARLQDLRLQDAVLGAAHPVPPGLPLFLPYYICVADEDDYRDFVNLDHAHSLLRDYQQREGIAMDQLLSQSLPNDGDEKYEKTIIKSGDQTFYKFMKRIAACQEQILRYSWSGEPLFLTCPTSEVTELPACSQCGGQRIFEFQLMPALVSMLKSANLGLSVEFGTILVYTCEKSCWPPNHQTPMEEFCIIQEDPDELLFK
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis
Primary or Secondary Primary
Gene ID (Entrez) 84306
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.