missing translation for 'onlineSavingsMsg'
Learn More

Calponin 2 Antibody (4C9) [Alexa Fluor« 405], Novus Biologicals Biologicals™

Product Code. 30518798 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30518798 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30518798 Supplier Novus Biologicals Supplier No. NBP342944AF405

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Calponin 2 Monoclonal antibody specifically detects Calponin 2 in Human,Porcine samples. It is validated for ELISA,Immunohistochemistry,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen Calponin 2
Applications ELISA, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 4C9
Conjugate Alexa Fluor 405
Formulation 50mM Sodium Borate
Gene Alias calponin 2, Calponin H2, smooth muscle, calponin-2, Neutral calponin
Host Species Mouse
Immunogen calponin 2 - (Uniprot# Q99439)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 1265
Target Species Human, Pig
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.