missing translation for 'onlineSavingsMsg'
Learn More

CTAGE1 Antibody (4F3) [DyLight 350], Novus Biologicals Biologicals™

Product Code. 30519005 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30519005 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30519005 Supplier Novus Biologicals Supplier No. NBP342950UV

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

CTAGE1 Monoclonal antibody specifically detects CTAGE1 in Human samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen CTAGE1
Applications ELISA, Immunohistochemistry, Western Blot, Immunocytochemistry/Immunofluorescence
Classification Monoclonal
Clone 4F3
Conjugate DyLight 350
Formulation 50mM Sodium Borate
Gene Alias Cancer/testis antigen 21.1, Cancer/testis antigen 21.2, CT21.1cancer/testis antigen family 21, member 1, CT21.2cancer/testis antigen family 21, member 2, CTAGE, CTAGE-1, cTAGE-2, CTAGE2, cutaneous T-cell lymphoma-associated antigen 1, cutaneous T-cell lymphoma-associated antigen 2, Protein cTAGE-1, protein cTAGE-2
Host Species Mouse
Immunogen cutaneous T-cell lymphoma-associated antigen 1 - (Uniprot# Q96RT6)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64693
Target Species Human
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.