missing translation for 'onlineSavingsMsg'
Learn More

17 beta-HSD14/HSD17B14 Antibody (6C4) [Janelia Fluor« 549], Novus Biologicals Biologicals™

Product Code. 30519992 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30519992 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30519992 Supplier Novus Biologicals Supplier No. NBP342967JF549

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

17 beta-HSD14/HSD17B14 Monoclonal antibody specifically detects 17 beta-HSD14/HSD17B14 in Human,Mouse samples. It is validated for ELISA,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen 17 beta-HSD14/HSD17B14
Applications ELISA, Western Blot
Classification Monoclonal
Clone 6C4
Conjugate Janelia Fluor 549
Formulation 50mM Sodium Borate
Gene Alias dehydrogenase/reductase (SDR family) member 10, Dehydrogenase/reductase SDR family member 10, DHRS1017-beta-hydroxysteroid dehydrogenase 14, EC 1.1.1.-, hydroxysteroid (17-beta) dehydrogenase 14,17-beta-hydroxysteroid dehydrogenase DHRS10, retinal short-chain dehydrogenase/reductase 3, Retinal short-chain dehydrogenase/reductase retSDR3, retSDR3, RETSDR3,17-beta-HSD 14, SDR3, SDR47C1, short chain dehydrogenase/reductase family 47C, member 1
Host Species Mouse
Immunogen hydroxysteroid(17-beta) dehydrogenase 10 - (Uniprot# Q99714 )
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 51171
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.