missing translation for 'onlineSavingsMsg'
Learn More

Monoamine Oxidase B Antibody (8E0) [PerCP], Novus Biologicals Biologicals™

Product Code. 30520778 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30520778 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30520778 Supplier Novus Biologicals Supplier No. NBP342993PCP

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Monoamine Oxidase B Monoclonal antibody specifically detects Monoamine Oxidase B in Human samples. It is validated for ELISA,Immunohistochemistry,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen Monoamine Oxidase B
Applications ELISA, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 8
Conjugate PerCP
Formulation PBS
Gene Alias adrenalin oxidase, amine oxidase [flavin-containing] B, EC 1.4.3, EC 1.4.3.4, MAO, brain, MAO, platelet, MAO-B, MGC26382, monoamine oxidase B, Monoamine oxidase type B, tyramine oxidase
Host Species Mouse
Immunogen monoamine oxidase B - (Uniprot# P27338)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Amino Acids Drugs and other small molecules, Cancer, Endocrinology, Neurodegeneration, Neuroscience, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 4129
Target Species Human
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.