missing translation for 'onlineSavingsMsg'
Learn More

MCRS1 Antibody (5G3) [DyLight 550], Novus Biologicals Biologicals™

Product Code. 30520840 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30520840 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30520840 Supplier Novus Biologicals Supplier No. NBP342995R

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

MCRS1 Monoclonal antibody specifically detects MCRS1 in Human,Mouse samples. It is validated for ELISA,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen MCRS1
Applications ELISA, Western Blot
Classification Monoclonal
Clone 5G3
Conjugate DyLight 550
Formulation 50mM Sodium Borate
Gene Alias Cell cycle-regulated factor p78, ICP22BP, INO80 complex subunit Q, INO80Q58 kDa microspherule protein, MCRS2INO80 complex subunit J, microspherule protein 1, MSP58cell cycle-regulated factor (78 kDa), P78
Host Species Mouse
Immunogen microspherule protein 1 - (Uniprot# Q96EZ8)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 10445
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.