missing translation for 'onlineSavingsMsg'
Learn More

MEST Antibody (5E0) [mFluor Violet 450 SE], Novus Biologicals Biologicals™

Product Code. 30520895 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30520895 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30520895 Supplier Novus Biologicals Supplier No. NBP342997MFV450

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

MEST Monoclonal antibody specifically detects MEST in Human,Mouse samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen MEST
Applications ELISA, Immunohistochemistry, Western Blot, Immunocytochemistry/Immunofluorescence
Classification Monoclonal
Clone 5
Conjugate mFluor Violet 450 SE
Formulation 50mM Sodium Borate
Gene Alias DKFZp686L18234, EC 3.-, mesoderm specific transcript (mouse) homolog, mesoderm specific transcript homolog (mouse), mesoderm-specific transcript homolog protein, MGC111102, MGC8703, paternally expressed gene 1, PEG1Paternally-expressed gene 1 protein
Host Species Mouse
Immunogen mesoderm specific transcript homolog(Mouse) - (Uniprot# Q5EB52)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Gene ID (Entrez) 4232
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.