missing translation for 'onlineSavingsMsg'
Learn More

SND1 Antibody (0A1) [DyLight 594], Novus Biologicals Biologicals™

Product Code. 30522083 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30522083 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30522083 Supplier Novus Biologicals Supplier No. NBP343037DL594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

SND1 Monoclonal antibody specifically detects SND1 in Human,Mouse samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen SND1
Applications ELISA, Immunohistochemistry, Western Blot, Immunocytochemistry/Immunofluorescence
Classification Monoclonal
Clone 0A1
Conjugate DyLight 594
Formulation 50mM Sodium Borate
Gene Alias 100 kDa coactivator, EBNA2 coactivator p100, P100, p100 co-activator, p100 EBNA2 co-activator, staphylococcal nuclease and tudor domain containing 1, staphylococcal nuclease domain containing 1, staphylococcal nuclease domain-containing protein 1, TDRD11EBNA-2 co-activator (100kD), Tudor domain-containing protein 11
Host Species Mouse
Immunogen staphylococcal nuclease and tudor domain containing 1 - (Uniprot# Q7KZF4)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 27044
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.