missing translation for 'onlineSavingsMsg'
Learn More

ST6 Sialyltransferase 6/ST6GALNAC6 Antibody (1F4) [DyLight 350], Novus Biologicals Biologicals™

Product Code. 30522189 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30522189 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30522189 Supplier Novus Biologicals Supplier No. NBP343040UV

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

ST6 Sialyltransferase 6/ST6GALNAC6 Monoclonal antibody specifically detects ST6 Sialyltransferase 6/ST6GALNAC6 in Human samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen ST6 Sialyltransferase 6/ST6GALNAC6
Applications ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 1F4
Conjugate DyLight 350
Formulation 50mM Sodium Borate
Gene Alias alpha-N-acetylgalactosaminide alpha-2,6-sialyltransferase 6, CMP-NeuAC:(beta)-N-acetylgalactosaminide (alpha)2,6-sialyltransferase member VI, EC 2.4.99, EC 2.4.99.-, EC 2.4.99.7, GalNAc alpha-2,6-sialyltransferase VI, hST6GalNAc VI, RP11-203J24.3, Sialyltransferase 7F, sialytransferase 7 ((alpha-N-acetylneuraminyl 2,3-betagalactosyl-1,3)-N-acetylgalactosaminide alpha-2,6-sialytransferase) F, SIAT7F, SIAT7-F, ST6 (alpha-N-acetyl-neuraminyl-2,3-beta-galactosyl-1,3)-N-acetylgalactosaminidealpha-2,6-sialyltransferase 6, ST6GalNAc VI, ST6GalNAcVI
Host Species Mouse
Immunogen ST6(alpha-N-acetyl-neuraminyl-2,3-beta-galactosyl-1,3)-N-acetylgalactosaminide alpha-2,6-sialyltransferase 6 - (Uniprot# Q969X2)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 30815
Target Species Human
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.