missing translation for 'onlineSavingsMsg'
Learn More

SLP-2 Antibody (5D10) [Biotin], Novus Biologicals Biologicals™

Product Code. 30522230 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30522230 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30522230 Supplier Novus Biologicals Supplier No. NBP343042B

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. Se returpolicy

Mouse Monoclonal Antibody

SLP-2 Monoclonal antibody specifically detects SLP-2 in Human,Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Immunoprecipitation,Western Blot
TRUSTED_SUSTAINABILITY

Tekniske data

Antigen SLP-2
Applications ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 5D10
Conjugate Biotin
Formulation PBS
Gene Alias EPB72-like protein 2, HSPC108, SLP2, SLP-2stomatin-like protein 2, stomatin (EPB72)-like 2, stomatin-like 2
Host Species Mouse
Immunogen stomatin(EPB72)-like 2 - (Uniprot# Q9UJZ1)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Core ESC Like Genes, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 30968
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG2a
Vis mere Vis mindre
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.