missing translation for 'onlineSavingsMsg'
Learn More

Ferritin mitochondrial Antibody (AMB8LK) [DyLight 488], Novus Biologicals Biologicals™

Product Code. 30522713 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30522713 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30522713 Supplier Novus Biologicals Supplier No. NBP343585G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Ferritin mitochondrial Monoclonal antibody specifically detects Ferritin mitochondrial in Human samples. It is validated for Immunoprecipitation
TRUSTED_SUSTAINABILITY

Specifications

Antigen Ferritin mitochondrial
Applications Immunoprecipitation
Classification Monoclonal
Clone AMB8LK
Conjugate DyLight 488
Formulation 50mM Sodium Borate
Gene Alias EC 1.16.3.1, ferritin H subunit, ferritin mitochondrial, ferritin, mitochondrial, mitochondrial ferritin, MTF
Host Species Mouse
Immunogen The original antibody was generated by immunizing female Balb/c mice with ferritin extracted from human spleen cells. (Uniprot# Q8N4E7)
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 94033
Target Species Human
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.