missing translation for 'onlineSavingsMsg'
Learn More

Troponin T Type 2 (cardiac) Antibody (CL13382) [Janelia Fluor™ 525], Novus Biologicals™

Product Code. 30525415 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30525415 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30525415 Supplier Novus Biologicals Supplier No. NBP343865JF525

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Troponin T Type 2 (cardiac) Monoclonal antibody specifically detects Troponin T Type 2 (cardiac) in Human samples. It is validated for Western Blot,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen Troponin T Type 2 (cardiac)
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone CL13382
Conjugate Janelia Fluor 525
Formulation 50mM Sodium Borate
Gene Alias cardiac, CMD1D, CMPD2, dilated 1D (autosomal dominant), hypertrophic 2, MGC3889, RCM3, TnTc, troponin T type 2 (cardiac), troponin T, cardiac muscle
Host Species Mouse
Immunogen This antibody was generated using a synthetic peptide of P45379, with the exact immunogen sequence remaining proprietary.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Biology, Cellular Markers
Primary or Secondary Primary
Gene ID (Entrez) 7139
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.