missing translation for 'onlineSavingsMsg'
Learn More

GPR17 Antibody (CL10474) [HRP], Novus Biologicals Biologicals™

Product Code. 30529272 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30529272 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30529272 Supplier Novus Biologicals Supplier No. NBP344216H

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

GPR17 Monoclonal antibody specifically detects GPR17 in Human samples. It is validated for Western Blot,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen GPR17
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone CL10474
Conjugate HRP
Formulation PBS
Gene Alias DKFZp686M18273, G protein-coupled receptor 17, G-protein coupled receptor 17, P2Y-like receptor, R12, UDP/CysLT receptor, uracil nucleotide/cysteinyl leukotriene receptor
Host Species Mouse
Immunogen This antibody was generated using a synthetic peptide of Q13304, with the exact immunogen sequence remaining proprietary.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline GPCR, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 2840
Target Species Human
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.