missing translation for 'onlineSavingsMsg'
Learn More

CTAG1B Antibody (CL10130) [Biotin], Novus Biologicals Biologicals™

Product Code. 30529535 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30529535 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30529535 Supplier Novus Biologicals Supplier No. NBP344225B

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

CTAG1B Monoclonal antibody specifically detects CTAG1B in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen CTAG1B
Applications Western Blot
Classification Monoclonal
Clone CL10130
Conjugate Biotin
Formulation PBS
Gene Alias Autoimmunogenic cancer/testis antigen NY-ESO-1, cancer antigen 3, cancer/testis antigen 1, cancer/testis antigen 1B, Cancer/testis antigen 6.1, CT6.1L antigen family member 2, CTAG1CTAG1A, ESO1CTAG, LAGE2, LAGE2A, LAGE2BLAGE-2, New York esophageal squamous cell carcinoma 1, NY-ESO-1
Host Species Mouse
Immunogen This antibody was generated using a synthetic peptide of P78358, with the exact immunogen sequence remaining proprietary.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1485
Target Species Human
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.