missing translation for 'onlineSavingsMsg'
Learn More

c-jun Antibody (CL9662) [DyLight 550], Novus Biologicals Biologicals™

Product Code. 30530843 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30530843 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30530843 Supplier Novus Biologicals Supplier No. NBP344268R

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

c-jun Monoclonal antibody specifically detects c-jun in Human samples. It is validated for Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen c-jun
Applications Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence
Classification Monoclonal
Clone CL9662
Conjugate DyLight 550
Formulation 50mM Sodium Borate
Gene Alias Activator protein 1, AP1, AP-1, c-Jun, enhancer-binding protein AP1, Jun activation domain binding protein, jun oncogene, jun proto-oncogene, Proto-oncogene c-Jun, transcription factor AP-1, V-jun avian sarcoma virus 17 oncogene homologp39, v-jun sarcoma virus 17 oncogene homolog, v-jun sarcoma virus 17 oncogene homolog (avian)
Host Species Mouse
Immunogen This antibody was generated using a synthetic peptide of P05412, with the exact immunogen sequence remaining proprietary.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Cell Cycle and Replication, Hypoxia, Immunology, Innate Immunity, Phospho Specific, Signal Transduction, Transcription Factors and Regulators, Tumor Suppressors, Wnt Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 3725
Target Species Human
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.