missing translation for 'onlineSavingsMsg'
Learn More

CD19 Antibody (CL15454) [CoraFluorÖ 1], Novus Biologicals Biologicals™

Product Code. 30531097 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30531097 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30531097 Supplier Novus Biologicals Supplier No. NBP344277CL1

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

CD19 Monoclonal antibody specifically detects CD19 in Human samples. It is validated for Western Blot,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD19
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone CL15454
Conjugate CoraFluor 1
Formulation PBS
Gene Alias B4, B-lymphocyte antigen CD19, B-lymphocyte surface antigen B4, CD19 antigen, CD19 molecule, CVID3, Differentiation antigen CD19, MGC12802, T-cell surface antigen Leu-12
Host Species Mouse
Immunogen This antibody was generated using a synthetic peptide of P15391, with the exact immunogen sequence remaining proprietary.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, B Cell Development and Differentiation Markers, Cancer Stem Cells, Cytokine Research, Immunology, Innate Immunity, Mesenchymal Stem Cell Markers, Myeloid derived Suppressor Cell, Signal Transduction, Stem Cell Lines, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 930
Target Species Human
Content And Storage Store at 4°C in the dark. Do not freeze.
Product Type Antibody
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.