missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LAG-3 Antibody (CL14765) [Alexa Fluor« 750], Novus Biologicals Biologicals™
Shop All Bio Techne ProductsDescription
LAG-3 Monoclonal antibody specifically detects LAG-3 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
Specifications
| Antigen | LAG-3 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | CL14765 |
| Conjugate | Alexa Fluor 750 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | CD223, CD223 antigen, LAG3, LAG-3, lymphocyte activating 3, lymphocyte activation gene 3 protein, lymphocyte-activation gene 3, Secreted lymphocyte activation gene 3 protein, sLAG-3 |
| Host Species | Mouse |
| Immunogen | This antibody was generated using a synthetic peptide of P18627, with the exact immunogen sequence remaining proprietary. |
| Purification Method | Protein A purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?