missing translation for 'onlineSavingsMsg'
Learn More

ELAVL2 Antibody (CL15323) [DyLight 680], Novus Biologicals Biologicals™

Product Code. 30532302 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30532302 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30532302 Supplier Novus Biologicals Supplier No. NBP344321FR

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

ELAVL2 Monoclonal antibody specifically detects ELAVL2 in Human,Mouse,Rat samples. It is validated for Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen ELAVL2
Applications Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence
Classification Monoclonal
Clone CL15323
Conjugate DyLight 680
Formulation 50mM Sodium Borate
Gene Alias ELAV (embryonic lethal, abnormal vision, Drosophila)-like 2 (Hu antigen B), ELAV-like neuronal protein 1, ELAV-like protein 2, HELN1, HEL-N1, Hu-antigen B, HuB, HUBELAV (embryonic lethal, abnormal vision, Drosophila)-like 2, Nervous system-specific RNA-binding protein Hel-N1
Host Species Mouse
Immunogen This antibody was generated using a synthetic peptide of Q12926, with the exact immunogen sequence remaining proprietary.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neuroscience, Vision
Primary or Secondary Primary
Gene ID (Entrez) 1993
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG3
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.