missing translation for 'onlineSavingsMsg'
Learn More

Tau Antibody (HL2982) - Azide and BSA Free, Novus Biologicals™

Product Code. 30537793 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30537793 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30537793 Supplier Novus Biologicals Supplier No. NBP345159

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Tau Monoclonal antibody specifically detects Tau in Canine,Human,Mouse,Rat samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen Tau
Applications Western Blot
Classification Monoclonal
Clone HL2982
Conjugate Unconjugated
Formulation PBS
Gene Alias DDPAC, FLJ31424, FTDP-17, G protein beta1/gamma2 subunit-interacting factor 1, MGC138549, microtubule-associated protein tau, MSTDMAPTL, MTBT1Neurofibrillary tangle protein, MTBT2, PHF-tau, PPND, tau, TAUPaired helical filament-tau
Host Species Rabbit
Immunogen Synthetic peptide of human Tau.
Purification Method Protein A purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Alzheimers Research, Apoptosis, Cell Biology, Cellular Markers, Chaperone Mediated Autophagy (CMA), Cytokine Research, MAP Kinase Signaling, Neurodegeneration, Neuroscience, Phospho Specific, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 4137
Target Species Canine, Human, Mouse, Rat
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.