missing translation for 'onlineSavingsMsg'
Learn More

PRPF6 Antibody - BSA Free, Novus Biologicals™

Product Code. 30556663 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30556663 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30556663 Supplier Novus Biologicals Supplier No. NBP346881

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

PRPF6 Polyclonal antibody specifically detects PRPF6 in Human,Mouse samples. It is validated for ELISA,Immunohistochemistry,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen PRPF6
Applications ELISA, Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000, ELISA, Immunohistochemistry 1:20-1:200
Formulation PBS, 50% Glycerol, pH (7.3)
Gene Alias androgen receptor N-terminal domain transactivating protein-1, ANT-1, bB152O15.1, C20orf14, chromosome 20 open reading frame 14, hPrp6, p102 U5 small nuclear ribonucleoprotein particle-binding protein, pre-mRNA-processing factor 6, Prp6, PRP6 homolog, PRP6 pre-mRNA processing factor 6 homolog (S. cerevisiae), PRP6 pre-mRNA processing factor 6 homolog (yeast), putative mitochondrial outer membrane protein import receptor, TOM, U5 snRNP-associated 102 kDa protein, U5-102 kDa protein, U5-102K
Host Species Rabbit
Immunogen PRP6 pre-mRNA processing factor 6 homolog. (Uniprot# O94906)
Purification Method Immunogen affinity purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 24148
Target Species Human, Mouse
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.