missing translation for 'onlineSavingsMsg'
Learn More

AP1M1 Antibody - BSA Free, Novus Biologicals™

Product Code. 30557157 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30557157 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30557157 Supplier Novus Biologicals Supplier No. NBP348136

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

AP1M1 Polyclonal antibody specifically detects AP1M1 in Human,Mouse,Rat samples. It is validated for ELISA,Immunoprecipitation,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen AP1M1
Applications ELISA, Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:200-1:2000, ELISA, Immunoprecipitation 1:200-1:2000
Formulation PBS, 50% Glycerol, pH (7.3)
Gene Alias Adaptor protein complex AP-1 mu-1 subunit, Adaptor-related protein complex 1 mu-1 subunit, adaptor-related protein complex 1, mu 1 subunit, AP47MU-1A, AP-mu chain family member mu1A, CLAPM2AP-1 complex subunit mu-1, clathrin adaptor protein AP47, Clathrin assembly protein complex 1 medium chain 1, clathrin assembly protein complex 1, medium chain, clathrin assembly protein complex AP1, mu subunit, Clathrin coat assembly protein AP47, Clathrin coat-associated protein AP47, CLTNM, golgi adaptor AP-1 47 kDa protein, Golgi adaptor HA1/AP1 adaptin mu-1 subunit, HA1 47 kDa subunit, Mu1A-adaptin, Mu-adaptin 1
Host Species Rabbit
Immunogen adaptor-related protein complex 1, mu 1 subunit. (Uniprot# Q9BXS5)
Purification Method Immunogen affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Immunology, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 8907
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.