missing translation for 'onlineSavingsMsg'
Learn More

ATP5G2 Antibody - BSA Free, Novus Biologicals™

Product Code. 30558717 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30558717 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30558717 Supplier Novus Biologicals Supplier No. NBP347922

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

ATP5G2 Polyclonal antibody specifically detects ATP5G2 in Human samples. It is validated for ELISA,Immunoprecipitation
TRUSTED_SUSTAINABILITY

Specifications

Antigen ATP5G2
Applications ELISA, Immunoprecipitation
Classification Polyclonal
Conjugate Unconjugated
Dilution ELISA, Immunoprecipitation 1:500-1:2000
Formulation PBS, 50% Glycerol, pH (7.3)
Gene Alias ATP synthase lipid-binding protein, mitochondrial, ATP synthase proteolipid P2, ATP synthase, H+ transporting, mitochondrial F0 complex, subunit c (subunit 9), ATP synthase, H+ transporting, mitochondrial F0 complex, subunit C2 (subunit 9), ATP synthase, H+ transporting, mitochondrial Fo complex, subunit C2 (subunit 9), ATPase protein 9, ATPase subunit c, isoform 2, mitochondrial ATP synthase, subunit C (subunit 9)
Host Species Rabbit
Immunogen ATP synthase, H+ transporting, mitochondrial F0 complex, subunit C2(subunit 9). (Uniprot# Q06055)
Purification Method Immunogen affinity purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 517
Target Species Human
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.