missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Archaemetzincin 2 Polyclonal antibody specifically detects Archaemetzincin 2 in Human,Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunocytochemistry/ Immunofluorescence
Specifications
Specifications
| Antigen | Archaemetzincin 2 |
| Applications | ELISA, Immunohistochemistry, Western Blot, Immunocytochemistry/Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:500-1:5000, ELISA, Immunohistochemistry 1:20-1:200, Immunocytochemistry/ Immunofluorescence 1:20-1:200 |
| Formulation | PBS, 50% Glycerol, pH (7.3) |
| Gene Alias | archaelysin family metallopeptidase 2, archaemetzincin-2, archaemetzincins-2, Archeobacterial metalloproteinase-like protein 2, EC 3.- |
| Host Species | Rabbit |
| Immunogen | archaelysin family metallopeptidase 2. (Uniprot# Q86W34) |
| Purification Method | Immunogen affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?