missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
NGX6 Polyclonal antibody specifically detects NGX6 in Human samples. It is validated for ELISA,Immunohistochemistry
Specifications
Specifications
| Antigen | NGX6 |
| Applications | ELISA, Immunohistochemistry |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | ELISA, Immunohistochemistry 1:20-1:200 |
| Formulation | PBS, 50% Glycerol, pH (7.3) |
| Gene Alias | C9orf127, chromosome 9 open reading frame 127, NAG-5, nasopharyngeal carcinoma expressed 6, nasopharyngeal carcinoma related protein, Nasopharyngeal carcinoma-associated gene 6 protein, NGX6MGC120460, Protein NAG-5, Protein NGX6, RP11-112J3.10, transmembrane protein 8B |
| Host Species | Rabbit |
| Immunogen | chromosome 9 open reading frame 127. (Uniprot# A6NDV4) |
| Purification Method | Immunogen affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?