missing translation for 'onlineSavingsMsg'
Learn More

SLC6A8 Antibody - BSA Free, Novus Biologicals™

Product Code. 30559657 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30559657 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30559657 Supplier Novus Biologicals Supplier No. NBP345900

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

SLC6A8 Polyclonal antibody specifically detects SLC6A8 in Human samples. It is validated for ELISA,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC6A8
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000, ELISA
Formulation PBS, 50% Glycerol, pH (7.3)
Gene Alias Creatine transporter 1, CRT, CRTRcreatine transporter SLC6A8, CT1MGC87396, sodium- and chloride-dependent creatine transporter 1, solute carrier family 6 (neurotransmitter transporter, creatine), member 8, Solute carrier family 6 member 8
Host Species Rabbit
Immunogen solute carrier family 6(neurotransmitter transporter, creatine), member 8. (Uniprot# P48029)
Purification Method Immunogen affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cancer, Endocrinology, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 6535
Target Species Human
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.