missing translation for 'onlineSavingsMsg'
Learn More

3alpha-Hydroxy-5beta-cholan-24-oic Acid (Lithocholic Acid), Mikromol™

Product Code. 17971686
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17971686 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17971686 Supplier Mikromol Supplier No. MM0835.030025
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

3alpha-Hydroxy-5beta-cholan-24-oic Acid (Lithocholic Acid)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Lithocholic Acid
CAS 434-13-9
Analyte or Component Names Lithocholic Acid
Shipping Condition Room Temperature
Molecular Formula C24 H40 O3
Certifications/Compliance ISO 9001
Synonym (3α,5β)-3-Hydroxycholan-24-oic acid, 17β-(1-Methyl-3-carboxypropyl)etiocholan-3α-ol, 3-Hydroxycholan-24-oic acid, 3α-Hydroxy-5β-cholan-24-oate, 3α-Hydroxy-5β-cholan-24-oic acid, 3α-Hydroxy-5β-cholanic acid, 3α-Hydroxy-5β-cholanoic acid, 3α-Hydroxycholanic acid, Lithocholic acid, NSC 683770, Chenodeoxycholic Acid Impurity C, Ursodeoxycholic Acid Impurity C
InChI Formula InChI=1S/C24H40O3/c1-15(4-9-22(26)27)19-7-8-20-18-6-5-16-14-17(25)10-12-23(16,2)21(18)11-13-24(19,20)3/h15-21,25H,4-14H2,1-3H3,(H,26,27)/t15-,16-,17-,18+,19-,20+,21+,23+,24-/m1/s1
SMILES C[C@H](CCC(=O)O)[C@H]1CC[C@H]2[C@@H]3CC[C@@H]4C[C@H](O)CC[C@]4(C)[C@H]3CC[C@]12C
IUPAC Name (4R)-4-[(3R,5R,8R,9S,10S,13R,14S,17R)-3-hydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentanoic acid
Molecular Weight (g/mol) 376.57
Quantity 25 mg
Formula Weight 376.2977 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.