missing translation for 'onlineSavingsMsg'
Learn More
Learn More
[(3R,3aS,6aR)-Hexahydrofuro[2,3-b]furan-3-yl] (2,5-Dioxopyrrolidin-1-yl) Carbonate, Mikromol™
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Description
[(3R,3aS,6aR)-Hexahydrofuro[2,3-b]furan-3-yl] (2,5-Dioxopyrrolidin-1-yl) Carbonate
Specifications
Specifications
| Chemical Name or Material | ((3R,3aS,6aR)-Hexahydrofuro[2,3-b]furan-3-yl)(2,5-Dioxopyrrolidin-1-yl) Carbonate |
| CAS | 253265-97-3 |
| Analyte or Component Names | ((3R,3aS,6aR)-Hexahydrofuro[2,3-b]furan-3-yl)(2,5-Dioxopyrrolidin-1-yl) Carbonate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C11 H13 N O7 |
| Certifications/Compliance | ISO 17025 |
| Synonym | Carbonic acid, 2,5-dioxo-1-pyrrolidinyl [(3R,3aS,6aR)-hexahydrofuro[2,3-b]furan-3-yl] ester, 1-[[[[(3R,3aS,6aR)-Hexahydrofuro[2,3-b]furan-3-yl]oxy]carbonyl]oxy]-5-pyrrolidinedione |
| InChI Formula | InChI=1S/C11H13NO7/c13-8-1-2-9(14)12(8)19-11(15)18-7-5-17-10-6(7)3-4-16-10/h6-7,10H,1-5H2/t6-,7-,10+/m0/s1 |
| SMILES | O=C(O[C@H]1CO[C@H]2OCC[C@@H]12)ON3C(=O)CCC3=O |
| IUPAC Name | [(3aS,4R,6aR)-2,3,3a,4,5,6a-hexahydrofuro[2,3-b]furan-4-yl] (2,5-dioxopyrrolidin-1-yl) carbonate |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction