missing translation for 'onlineSavingsMsg'
Learn More

(3R,5R)-7-[(1S,2S,6R,8S,8aR)-8-[(2,2-Dimethylbutanoyl)oxy]-2,6-dimethyl-1,2,6,7,8,8a-hexahydronaphthalen-1-yl]-3,5-dihydroxyheptanoic Acid Sodium Salt, Mikromol™

Product Code. 17902816
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17902816 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17902816 Supplier Mikromol Supplier No. MM0158.01
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

(3R,5R)-7-[(1S,2S,6R,8S,8aR)-8-[(2,2-Dimethylbutanoyl)oxy]-2,6-dimethyl-1,2,6,7,8,8a-hexahydronaphthalen-1-yl]-3,5-dihydroxyheptanoic Acid Sodium Salt
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (3R,5R)-7-[(1S,2S,6R,8S,8aR)-8-[(2,2-Dimethylbutanoyl)oxy]-2,6-dimethyl-1,2,6,7,8,8a-hexahydronaphthalen-1-yl]-3,5-dihydroxyheptanoic Acid Sodium Salt
CAS 101314-97-0
Analyte or Component Names (3R,5R)-7-[(1S,2S,6R,8S,8aR)-8-[(2,2-Dimethylbutanoyl)oxy]-2,6-dimethyl-1,2,6,7,8,8a-hexahydronaphthalen-1-yl]-3,5-dihydroxyheptanoic Acid Sodium Salt
Shipping Condition Ice pack (-20°C)
Molecular Formula C25 H39 O6 . Na
Certifications/Compliance ISO 9001
Synonym Simvastatin Imp. A (EP), (3R,5R)-7-[(1S,2S,6R,8S,8aR)-8-[(2,2-Dimethylbutanoyl)oxy]-2,6-dimethyl-1,2,6,7,8,8a-hexahydronaphthalen-1-yl]-3,5-dihydroxyheptanoic Acid Sodium Salt, Simvastatin Imp. A (EP) as Sodium Salt, Tenivastatin Sodium Salt, Simvastatin Hydroxy Acid Sodium Salt, Simvastatin Impurity A as Sodium Salt
InChI Formula InChI=1S/C25H40O6.Na/c1-6-25(4,5)24(30)31-21-12-15(2)11-17-8-7-16(3)20(23(17)21)10-9-18(26)13-19(27)14-22(28)29;/h7-8,11,15-16,18-21,23,26-27H,6,9-10,12-14H2,1-5H3,(H,28,29);/q;+1/p-1/t15-,16-,18+,19+,20-,21-,23-;/m0./s1
SMILES [Na+].CCC(C)(C)C(=O)O[C@H]1C[C@@H](C)C=C2C=C[C@H](C)[C@H](CC[C@@H](O)C[C@@H](O)CC(=O)[O-])[C@@H]12
IUPAC Name sodium;(3R,5R)-7-[(1S,2S,6R,8S,8aR)-8-(2,2-dimethylbutanoyloxy)-2,6-dimethyl-1,2,6,7,8,8a-hexahydronaphthalen-1-yl]-3,5-dihydroxyheptanoate
Molecular Weight (g/mol) 458.56
Quantity 100 mg
Formula Weight 458.2644 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.