missing translation for 'onlineSavingsMsg'
Learn More

4-(1H-1,2,4-Triazol-1-ylmethyl)benzonitrile, Mikromol

Product Code. 30459312
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30459312 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30459312 Supplier Mikromol Supplier No. MM1143.030025
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol- your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

4-(1H-1,2,4-Triazol-1-ylmethyl)benzonitrile, Mikromol
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-(1H-1,2,4-Triazol-1-ylmethyl)benzonitrile
CAS 112809-25-3
Analyte or Component Names 4-(1H-1,2,4-Triazol-1-ylmethyl)benzonitrile
Shipping Condition Room Temperature
Molecular Formula C10 H8 N4
Certifications/Compliance ISO 9001
Synonym Benzonitrile, 4-(1H-1,2,4-triazol-1-ylmethyl)-, 4-(1H-1,2,4-Triazol-1-ylmethyl)benzonitrile, 1-(4-Cyanobenzyl)-1,2,4-triazole, 4-1-(1,2,4-Triazolyl)-methyl-benzonitrile, 4-2[1-(1,2,4-Triazol-1-yl)methyl]benzonitrile, 4-[(1,2,4-Triazol-1-yl)methyl]benzonitrile, 4-[(1H-1,2,4-Triazol-1-yl)methyl]benzonitrile
InChI Formula InChI=1S/C10H8N4/c11-5-9-1-3-10(4-2-9)6-14-8-12-7-13-14/h1-4,7-8H,6H2
Solution Type Neat
SMILES N#Cc1ccc(Cn2cncn2)cc1
IUPAC Name 4-(1,2,4-triazol-1-ylmethyl)benzonitrile
Molecular Weight (g/mol) 184.2
Quantity 25 mg
Formula Weight 184.0749 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.