missing translation for 'onlineSavingsMsg'
Learn More

4-(2-Aminoethyl)-2-methoxyphenol Hydrochloride, Mikromol™

Product Code. 17932586
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17932586 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17932586 Supplier Mikromol Supplier No. MM0384.030025
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

4-(2-Aminoethyl)-2-methoxyphenol Hydrochloride
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3-Methoxy-p-tyramine Hydrochloride
CAS 1477-68-5
Analyte or Component Names 3-Methoxy-p-tyramine Hydrochloride
Shipping Condition Room Temperature
Molecular Formula C9 H13 N O2 . Cl H
Certifications/Compliance ISO 9001
Synonym 3-O-Methyldopamine Hydrochloride, 3-Methoxy Tyramine HCl
InChI Formula InChI=1S/C9H13NO2.ClH/c1-12-9-6-7(4-5-10)2-3-8(9)11;/h2-3,6,11H,4-5,10H2,1H3;1H
SMILES Cl.COc1cc(CCN)ccc1O
IUPAC Name 4-(2-aminoethyl)-2-methoxyphenol;hydrochloride
Molecular Weight (g/mol) 203.67
Quantity 25 mg
Formula Weight 203.0713 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.