missing translation for 'onlineSavingsMsg'
Learn More

4-[2-[(Cyclohexylcarbamoyl)amino]ethyl]benzenesulphonamide, Mikromol™

Product Code. 30051413
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30051413 25 mg 25mg
30049185 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30051413 Supplier Mikromol Supplier No. MM0942.080025
 Restricted Item
Estimated Shipment: 03-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

4-[2-[(Cyclohexylcarbamoyl)amino]ethyl]benzenesulphonamide
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-[2-[(Cyclohexylcarbamoyl)amino]ethyl]benzenesulphonamide
CAS 10080-05-4
Analyte or Component Names 4-[2-[(Cyclohexylcarbamoyl)amino]ethyl]benzenesulphonamide
Shipping Condition Room Temperature
Molecular Formula C15 H23 N3 O3 S
Certifications/Compliance ISO 17025
Synonym 4-[2-[(Cyclohexylcarbamoyl)amino]ethyl]benzenesulphonamide, Glipizide Imp. H (EP), Glipizide Impurity H
InChI Formula InChI=1S/C15H23N3O3S/c16-22(20,21)14-8-6-12(7-9-14)10-11-17-15(19)18-13-4-2-1-3-5-13/h6-9,13H,1-5,10-11H2,(H2,16,20,21)(H2,17,18,19)
SMILES NS(=O)(=O)c1ccc(CCNC(=O)NC2CCCCC2)cc1
IUPAC Name 1-cyclohexyl-3-[2-(4-sulfamoylphenyl)ethyl]urea
Molecular Weight (g/mol) 325.43
Quantity 25 mg
Formula Weight 325.146 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.