missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-[(2-Methoxyphenoxy)methyl]-1,3-dioxolan-2-one, Mikromol™
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Description
4-[(2-Methoxyphenoxy)methyl]-1,3-dioxolan-2-one
Specifications
Specifications
| Chemical Name or Material | rac Guaifenesin Cyclic Carbonate |
| CAS | 2049-21-0 |
| Analyte or Component Names | rac Guaifenesin Cyclic Carbonate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C11 H12 O5 |
| Certifications/Compliance | ISO 9001 |
| Synonym | 1,3-Dioxolan-2-one, 4-[(2-methoxyphenoxy)methyl]-, Carbonic acid, cyclic [(o-methoxyphenoxy)methyl]ethylene ester (6CI,7CI,8CI), 1,2-Propanediol, 3-(o-methoxyphenoxy)-, cyclic carbonate (8CI), 4-[(2-Methoxyphenoxy)methyl]-1,3-dioxolan-2-one |
| InChI Formula | InChI=1S/C11H12O5/c1-13-9-4-2-3-5-10(9)14-6-8-7-15-11(12)16-8/h2-5,8H,6-7H2,1H3 |
| SMILES | COc1ccccc1OCC2COC(=O)O2 |
| IUPAC Name | 4-[(2-methoxyphenoxy)methyl]-1,3-dioxolan-2-one |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction