missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4′-[[4-Methyl-6-(1-methyl-1H-benzimidazol-2-yl)-2-propyl-1H-benzimidazol-1-yl]methyl][1,1′-biphenyl]-2-carboxamide (Telmisartan Amide), Mikromol™
Description
4′-[[4-Methyl-6-(1-methyl-1H-benzimidazol-2-yl)-2-propyl-1H-benzimidazol-1-yl]methyl][1,1′-biphenyl]-2-carboxamide (Telmisartan Amide)
Specifications
Specifications
| Chemical Name or Material | Telmisartan Amide |
| CAS | 915124-86-6 |
| Analyte or Component Names | Telmisartan Amide |
| Shipping Condition | Room Temperature |
| Molecular Formula | C33 H31 N5 O |
| Certifications/Compliance | ISO 9001 |
| Synonym | 4'-[[4-Methyl-6-(1-methyl-1H-benz-imidazol-2-yl)-2-propyl-1H-benzimidazol-1-yl]-methyl]biphenyl-2-carboxamide, 4'-[(1,4'-Dimethyl-2'-propyl[2,6'-bi-1H-benzimidazol]-1'-yl)methyl][1,1'-biphenyl]-2-carboxamide, [1,1'-Biphenyl]-2-carboxamide, 4'-[(1,4'-dimethyl-2'-propyl[2,6'-bi-1H-benzimidazol]-1'-yl)methyl]-, Telmisartan Imp F (EP), Telmisartan Impurity F |
| InChI Formula | InChI=1S/C33H31N5O/c1-4-9-30-36-31-21(2)18-24(33-35-27-12-7-8-13-28(27)37(33)3)19-29(31)38(30)20-22-14-16-23(17-15-22)25-10-5-6-11-26(25)32(34)39/h5-8,10-19H,4,9,20H2,1-3H3,(H2,34,39) |
| SMILES | CCCc1nc2c(C)cc(cc2n1Cc3ccc(cc3)c4ccccc4C(=O)N)c5nc6ccccc6n5C |
| IUPAC Name | 2-[4-[[4-methyl-6-(1-methylbenzimidazol-2-yl)-2-propylbenzimidazol-1-yl]methyl]phenyl]benzamide |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?